Learn More
Perkin Elmer US LLC ICP-MS Alternate Internal Standard 1, 5-10 %, HNO3, 500 mL
Catalog No. NC1626672
Encompass_Preferred
Concentration of Element includes: Matrix Concentration 5-10 %, HNO3 10 µg/mL: Bi, Ge, In, Li6, Sc, Tb, Y
Catalog No. NC1626672 Supplier Perkin Elmer US LLC Supplier No. N9303833
Concentration of Element includes: Matrix Concentration 5-10 %, HNO3 10 µg/mL: Bi, Ge, In, Li6, Sc, Tb, Y