Learn More
Internal Standard Solution (BDMC) (For EPA 531.1)
Catalog No. USPPS1801
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
Catalog No. Quantity
US-PPS-1801 1 x 1 mL
1 options
Catalog No. US-PPS-1801 Supplier Agilent Technologies Supplier No. PPS1801
4-Bromo-3,5-dimethylphenyl-N-methylcarbamate (BDMC) at 100μg/mL in methanol
Specifications
Chemical Name or Material Internal Standard Solution (BDMC)
Packaging Ampule
Quantity 1 x 1 mL