Learn More
Internal Standard Solution (DBOB) (For EPA 515.1)
Catalog No. USPPS1701
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
Catalog No. Quantity
US-PPS-1701 1 x 1 mL
1 options
Catalog No. US-PPS-1701 Supplier Agilent Technologies Supplier No. PPS1701
4,4'-Dibromooctafluorobiphenyl at 100μg/mL in Methyl tert-Butyl Ether
Specifications
Chemical Name or Material Internal Standard Solution (DBOB)
Packaging Ampule
Quantity 1 x 1 mL