Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Internal Standard Solution (For EPA 507)

Catalog No. USPPS1101
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-PPS-1101 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-PPS-1101 Supplier Agilent Technologies Supplier No. PPS1101
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Triphenyl Phosphate (TPP) at 500μg/mL in Methyl-tert Butyl Ether

Chemical Identifiers

CAS 1634-04-4
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
InChI Key BZLVMXJERCGZMT-UHFFFAOYSA-N
PubChem CID 15413
ChEBI CHEBI:27642
IUPAC Name 2-methoxy-2-methylpropane
SMILES CC(C)(C)OC

Specifications

Packaging Ampule
Quantity 1 x 1 mL
Chemical Name or Material Internal Standard Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.