Learn More
Perkin Elmer US LLC Internal Standards Kit for MEINHARD Plus Nebulizer
Catalog No. 502111335
Encompass_Preferred
NexION 1000/ 2000 internal standards kits utilize a low-internal volume mixing tee for in-line addition of internal standard with sample. Utilizing the MP2 micro peristaltic pump to achieve the highest level of accuracy and precision, the internal standards kit is available for both PFA-ST and MEINHARD Plus nebulizers.
Catalog No. 50-211-1335 Supplier Perkin Elmer US LLC Supplier No. N8152421
NexION 1000/ 2000 internal standards kits utilize a low-internal volume mixing tee for in-line addition of internal standard with sample. Utilizing the MP2 micro peristaltic pump to achieve the highest level of accuracy and precision, the internal standards kit is available for both PFA-ST and MEINHARD Plus nebulizers.