Learn More
Inorganic Ventures 200.8 INTERNAL STD SOLN 125mL
SDP
Catalog No. 50723005
Encompass_Preferred
Matrix: Nitric Acid (HNO3), Dilution: 1:100 to 1:1000, Analyte: Bi, Sc, Y, In, and Tb 20 ug/mL each.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-005 Supplier Inorganic Ventures Supplier No. 2008ISS125ML
Matrix: Nitric Acid (HNO3), Dilution: 1:100 to 1:1000, Analyte: Bi, Sc, Y, In, and Tb 20 ug/mL each.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search