Learn More
Inorganic Ventures 6020CLPM9 INT STD SOLN 125mL
SDP
Catalog No. 50723017
Encompass_Preferred
Matrix: Nitric Acid (HNO3), Dilution: 1:100, Analyte: Bi, In, Rh, Tb, Ho, 6Li, Sc, and Y 10 ug/mL each.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-017 Supplier Inorganic Ventures Supplier No. 6020ISS125ML
Matrix: Nitric Acid (HNO3), Dilution: 1:100, Analyte: Bi, In, Rh, Tb, Ho, 6Li, Sc, and Y 10 ug/mL each.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search