Learn More
International Enviroguard™ Chemsplash™ 1 Coveralls
Catalog No. 19472685 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Size:
2X-Large
4X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-472-685 4X-Large
19-472-683 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-472-685 Supplier International Enviroguard™ Supplier No. 7019YT4XL
Add to Cart
Add to Cart
This item is not returnable. View return policy
Chemsplash™ 1 Coveralls offer breathable impervious protection.
Combines the economy of microporous technology with a revolutionary process of creating a surface augmented with additional polymers resulting in a barrier, which retains breathability for comfort while resisting many chemicals found in the workplace.
  • Chemsplash 1 is tested for chemical penetration and permeation resistance by exposure to accidental liquid splash
  • Seams are stitched and then taped to provide added safety from incidental chemical splash
Specifications
Size 4X-Large
Material Laminated Non-woven Fabric
Color Yellow
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached
Closure Type Front Zipper
Seam Type Taped
Sterility Non-sterile
Resistance Bloodborne Pathogens, Chemicals
Back Style Elastic