Learn More
International Enviroguard™ GammaGuard CE™ Clean Processed Sterile Boot Covers
Catalog No. 19472645 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Size:
Standard
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-472-645 X-Large
19-472-644 Standard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-472-645 Supplier International Enviroguard™ Supplier No. CE11106CISXL
Add to Cart
Add to Cart
This item is not returnable. View return policy
Low-linting, sterile cleanroom boot covers are made with soft and breathable microporous fabric. Each pair is individually packaged to help prevent cross-contamination.
Sterile boot covers are made of comfortable fabric and include non-skid soles to help prevent accidental slips. Boot covers provide protection from liquids and particulates. Each boot cover is low-linting and sterilized to a 10-6 sterility assurance level (SAL). The fabric helps reduce the release of unwanted particulates in the work area while tunnelized elastic provides a more secure fit. This eliminates raw edges that may also generate unwanted particulates. Each pair of boot covers is individually packaged to help prevent cross-contamination.
  • Sterile-Don™ packaging protects against contamination during donning
  • GammaGuard CE™ helps keep wearers cooler vs many competitive fabrics
  • Tear-resistant fabric ensures your safety in demanding environments
  • Meets high manufacturing standards - made in an ISO-certified facility
  • Certificate of sterility included in each case
  • Bound seams
Order Info
Sterile-Don™ Packaging - Certificate of Sterility included in each case.
Specifications
Size X-Large
Material Microporous Fabric
Latex Free Yes
Color White
Sterility Sterile
Closure Type Elastic Top
Sole Type Non-skid
Seam Type Bound
Thickness 34 mil
Certifications/Compliance ISO, ANSI ISEA 101-1996 R2034
Packaging Quantity 200/Cs.
Packaging Type Individually Packaged Pairs
Height (English) 18 in.
Height (Metric) 45.72 cm
Product Type Controlled Environments Boot Covers