Learn More
International Enviroguard™ GammaGuard CE™ Sterile Cleanroom Hoods
Catalog No. 19472641 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Quantity:
100/Cs.
Catalog No. Quantity
19-472-641 100/Cs.
1 options
This item is not returnable. View return policy
Catalog No. 19-472-641 Supplier International Enviroguard™ Supplier No. CE11063CIS
Add to Cart
Add to Cart
This item is not returnable. View return policy
GammaGuard CE™ Sterile Cleanroom Hoods are made of a comfortable fabric that protects against various liquids and particulates.
  • Sterile
  • Each hood is individually packaged and sterilized to a 10-6 sterility assurance level (SAL)
  • Low-linting fabric helps reduce the release of unwanted particulates
  • Designed with tunnelized elastic face providing a more secure closure and stronger seams
  • Hoods include ties in the back to keep strings out of the way
  • GammaGuard CE™ fabric helps keep the wearer cooler
  • Tear-resistant fabric ensures your safety in challenging environments
  • Made in an ISO-certified facility
Specifications
Size Universal
Material Low Linting Fabric
Color White
Closure Type Ties in Back
Design Type Tunnelized Elastic Face
Latex Free Yes
Model CE11063CIS
Seam Type Serged
Sterility Sterile
Packaging Type Individually Packaged
Quantity 100/Cs.
Resistance Liquids