Learn More
International Enviroguard™ GammaGuard CE™ Sterile Cleanroom Shoe Covers
Catalog No. 19472642 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Size:
Standard
X-Large
Thickness:
31 mil
32 mil
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Thickness
19-472-642 Standard 31 mil
19-472-643 X-Large 32 mil
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-472-642 Supplier International Enviroguard™ Supplier No. CE11101CISL
Add to Cart
Add to Cart
This item is not returnable. View return policy
Disposable sterile cleanroom shoe covers are made from high MVTR, innovative fabric. Each pair is individually packaged to help prevent cross-contamination.
Sterile shoe covers are made of comfortable fabric and include non-skid soles to help prevent accidental slips as you work. Shoe covers protect against liquids and particulates, so your shoes stay pristine. Each shoe cover is low-linting and sterilized to the highest Sterility Assurance Level (SAL) of 10-6. The low-linting fabric helps reduce the release of unwanted particulates in your work area while tunnelized elastic also contains particle shed. Each pair of shoe covers is individually bagged to help prevent cross-contamination. Each bag includes a red indicator dot for proof of sterilization.
  • Sterile-Don™ packaging protects against contamination during donning
  • GammaGuard CE™ helps keep wearers cooler vs many competitive fabrics
  • Tear-resistant fabric ensures your safety in demanding environments
  • Meets high manufacturing standards - made in an ISO-certified facility
  • Certificate of sterility included in each case
  • Bound seams
Specifications
Size Standard
Material Microporous Fabric
Latex Free Yes
Color White
Disposable Single-use
Sterility Sterile
Closure Type Elastic Ankle
Sole Type Non-skid
Seam Type Bound
Thickness 31 mil
Certifications/Compliance ISO, ANSI ISEA 101-1996 R2034
Packaging Quantity 200 Pairs/Cs.
Packaging Type Individually Packaged
Height (English) 18 in.
Height (Metric) 45.72 cm
Product Type Controlled Environments Shoe Covers