Learn More
International Enviroguard™ MicroGuard CE™ Clean Processed Microporous Boot Covers
Catalog No. 19472560 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Size:
Large
X-Large
Thickness:
25 mil
27 mil
28 mil
29 mil
30 mil
Packaging Type:
Bulk Packaged
Individually Packaged
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Thickness Packaging Type
19-472-560 X-Large 30 mil Individually Packaged
19-472-558 Large 28 mil Individually Packaged
19-472-557 Large 27 mil Bulk Packaged
19-472-555 Large 25 mil Bulk Packaged
19-472-559 X-Large 29 mil Bulk Packaged
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-472-560 Supplier International Enviroguard™ Supplier No. CE8106CIXL
Add to Cart
Add to Cart
This item is not returnable. View return policy
Disposable boot covers are made from high MVTR, innovative fabric. Ideal for ISO 5 or higher non-hazardous work environments where reliable liquid and particulate protection is needed.
These microporous boot covers are clean processed to offer the perfect balance of safety and comfort. The fabric keeps dust and liquids out while allowing air to flow freely, so your feet stay cool throughout a hard day's work. These disposable boot covers are ideal for non-hazardous work environments, such as controlled environments or cleanrooms, where you need reliable liquid and particulate protection. Boot covers are designed with tunnelized elastic to contain particle shed. Non-skid soles provide added traction to help prevent accidental slips and trips. Each boot cover is individually packaged to help prevent cross-contamination.
  • Non-skid soles help improve traction to prevent accidental slips
  • Elastic closures for snug, secure fit that keeps boot covers in place
  • Available in Bulk or Individually packaged to prevent cross-contamination
Specifications
Size X-Large
Material Microporous Fabric
Color White
Closure Type Elastic Top and Ankles
Sole Type Non-skid
Seam Type Serged
Thickness 30 mil
Certifications/Compliance ANSI ISEA 101-1996 R2020, ASTM T903, ASTM F1670-07
Packaging Quantity 200/Cs.
Packaging Type Individually Packaged
Height (English) 17 in.
Height (Metric) 43.18 cm
Product Type Controlled Environments Boot Covers