Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

International Enviroguard™ MicroGuard CE™ Cleanroom Frocks

Catalog No. 19472544 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Packaging Type:
Bulk Packaged
Individually Packaged
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Packaging Type
19-472-544 Large Individually Packaged
19-472-543 Medium Individually Packaged
19-472-545 X-Large Individually Packaged
19-472-547 3X-Large Individually Packaged
19-472-548 4X-Large Individually Packaged
19-472-546 2X-Large Individually Packaged
19-472-537 Medium Bulk Packaged
19-472-541 3X-Large Bulk Packaged
19-472-538 Large Bulk Packaged
19-472-539 X-Large Bulk Packaged
19-472-542 4X-Large Bulk Packaged
19-472-540 2X-Large Bulk Packaged
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Catalog No. 19-472-544 Supplier International Enviroguard™ Supplier No. CE8046CIL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cleanroom frocks utilize state-of-the-art lamination technology to combine a soft non-woven fabric with a microporous film.

  • Microporous, clean processed frocks available in individual or bulk packaging
  • Fabric keeps dust and liquids out while allowing air to flow freely
  • Ideal for non-hazardous work environments where reliable liquid and particulate protection is needed
  • Designed with tunnelized elastic to contain particle shed
  • Bound mandarin collar for enhanced protection
  • Front snap closures are easy to secure
  • Constructed with bound seams to eliminate raw edges and strengthen the seam
  • No pockets

Specifications

Size Large
Material Microporous Fabric
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Snaps
Cuff Style Elastic
Collar Type Bound Mandarin
Size (English) Chest 40 in.
Size (Metric) Chest 106.6 cm
Certifications/Compliance CE
Packaging Type Individually Packaged
Length (Metric) 111.7 cm
Weight (English) 2.1 oz.
Weight (Metric) 0.05 kg
Cleanroom Class Class 100 (ISO 5)
Length (English) 44 in.
Product Type Controlled Environments Frock
Seam Type Bound
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.