Learn More
International Enviroguard™ MicroGuard CE™ Cleanroom Sleeves
Catalog No. 19472562 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Packaging:
Bulk Packaged
Individually Packaged
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
19-472-562 Individually Packaged
19-472-561 Bulk Packaged
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-472-562 Supplier International Enviroguard™ Supplier No. CE8065CI
Add to Cart
Add to Cart
This item is not returnable. View return policy
MicroGuard CE™ Cleanroom Sleeves utilize state-of-the-art lamination technology to combine a soft non-woven fabric with a microporous film.
  • Microporous, clean processed sleeves
  • Fabric keeps dust and liquids out while allowing air to flow freely
  • Ideal for nonhazardous work environments to protect against liquid and particulates
  • Designed with tunnelized elastic to offer a more secure closure, stronger seams, and the elimination of raw edges that may generate unwanted particulates
  • Each sleeve is individually packaged to help prevent cross-contamination
  • Elastic closures on both ends of sleeves for secure fit
  • Environmentally friendly - can be recycled after use
  • 18 in. length provides extended arm coverage
Order Info
Available in bulk packages or individually wrapped clean process packages
Specifications
Product Type Controlled Environment Sleeve
Size Standard
Material Microporous Fabric
Sterility Non-sterile
Disposable Single-use
Cuff Style Elastic
Model CE8065
Length (English) 18 in.
Length (Metric) 45.7 cm
Packaging Individually Packaged