Learn More
Fritsch Milling And Sizing Inc Interposed Sieve Pan 200/50mm Stl. Steel
SDP
Catalog No. 501534177
Encompass
For dry sieving with test sieves 200 mm dia.
If several sieving operations should be carried out simultaneously, an interposed sieve pan must be used as sieve pan between the sieves.
Catalog No. 50-153-4177 Supplier Fritsch Milling And Sizing Inc Supplier No. 31.1300.03
For dry sieving with test sieves 200 mm dia.
If several sieving operations should be carried out simultaneously, an interposed sieve pan must be used as sieve pan between the sieves.