Learn More
Fritsch Milling And Sizing Inc Interposed Sieve Pan 400/65 mm Stl. Steel
SDP
Catalog No. 501534350
Encompass
For dry sieving with test sieves 400 mm diameter.
If several sieving operations should be carried out simultaneously, an interposed sieve pan must be used as sieve pan between the sieves.
Catalog No. 50-153-4350 Supplier Fritsch Milling And Sizing Inc Supplier No. 37.1100.01
For dry sieving with test sieves 400 mm diameter.
If several sieving operations should be carried out simultaneously, an interposed sieve pan must be used as sieve pan between the sieves.