Learn More
Fritsch Milling And Sizing Inc Interposed Sieve Pan 8/2"
SDP
Supplier:  Fritsch Milling And Sizing Inc 31.1320.03
Encompass
For dry sieving with test sieves 8" dia.
If several sieving operations should be carried out simultaneously, an interposed sieve pan must be used as sieve pan between the sieves.
Catalog No. 50-153-4178