Learn More
Fritsch Milling And Sizing Inc Interposed Sieving Ring 8"
SDP
Catalog No. 501534164
Encompass
For wet sieving with test sieves 8" dia.
For simultaneous spraying the sieves above and below.
Catalog No. 50-153-4164 Supplier Fritsch Milling And Sizing Inc Supplier No. 31.0250.00
For wet sieving with test sieves 8" dia.
For simultaneous spraying the sieves above and below.