Learn More
GL Sciences Inc Inertsil C8 Analytical Column, 5um, 4.6x150mm
Catalog No. NC1772152
Encompass
Inertsil C8 columns have a pore size of 150 Å and it is recommended for rapid analysis of highly hydrophobic compounds. We recommend InertSustain C8 or Inertsil C8-4 columns for all new method development.
Catalog No. NC1772152 Supplier GL Sciences Inc Supplier No. 502001224
Inertsil C8 columns have a pore size of 150 Å and it is recommended for rapid analysis of highly hydrophobic compounds. We recommend InertSustain C8 or Inertsil C8-4 columns for all new method development.