Learn More
GL Sciences Inc Inertsil ODS-3 Analytical Column, 3µm, 150 x 4.6mm
Catalog No. NC1725527
Encompass
Inertsil ODS-3 is still GL Sciences’ most popular phase and continues to be used world widely and reliably for long established methods in pharmaceutical, and contract research labs. Inertsil ODS-3 has a relatively strong retentivity compared to commercially available ODS columns. In addition, the introduction of higher surface area silica provide high preparative loading capacity without sacrificing peak shape.
Catalog No. NC1725527 Supplier GL Sciences Inc Supplier No. 502001771
Inertsil ODS-3 is still GL Sciences’ most popular phase and continues to be used world widely and reliably for long established methods in pharmaceutical, and contract research labs. Inertsil ODS-3 has a relatively strong retentivity compared to commercially available ODS columns. In addition, the introduction of higher surface area silica provide high preparative loading capacity without sacrificing peak shape.