Learn More
GL Sciences Inc Inertsil ODS-3 Replacement Cartridge E, 5um, 4x10mm
Catalog No. NC1770401
Encompass
Inertsil ODS-3 replacement cartriage. Partible size: 5um | I.D.: 4.0mm | Length: 10mm | Applicable Column's I.D.: 4.0, 4.6mm
Catalog No. NC1770401 Supplier GL Sciences Inc Supplier No. 502019003
Inertsil ODS-3 replacement cartriage. Partible size: 5um | I.D.: 4.0mm | Length: 10mm | Applicable Column's I.D.: 4.0, 4.6mm