Learn More
MSC Intertape Double-Sided Tape, Polyester/Acrylic, 1.5 in. W, 55 m
Catalog No. 5000674495
Encompass
This double-sided tape features a polyester film construction with a solvent-based acrylic adhesive and a polyester film liner. It is designed for permanent applications, including core starting, splicing paper and film, mounting, and other general-purpose tasks requiring the secure joining of two surfaces. The tape is clear, 1.5 in. (38.10 mm) wide, and 0.0094 in. (3.2000 mil) thick.
  • Bonds to almost any surface
  • Not repulpable
Catalog No. 50-006-74495 Supplier MSC Supplier No. 72876121
This double-sided tape features a polyester film construction with a solvent-based acrylic adhesive and a polyester film liner. It is designed for permanent applications, including core starting, splicing paper and film, mounting, and other general-purpose tasks requiring the secure joining of two surfaces. The tape is clear, 1.5 in. (38.10 mm) wide, and 0.0094 in. (3.2000 mil) thick.
  • Bonds to almost any surface
  • Not repulpable