Learn More
MSC Intertape Masking Tape, 18 mm, 72 yd, Green, 1 ea
Catalog No. 5000792568
Encompass
This masking tape is crafted from polyester film with a silicone adhesive, providing excellent performance for various applications. It measures 18 mm wide, 72 yd long, and 3.4 mil thick, featuring a distinctive green color.
  • Made from polyester film
  • Features silicone adhesive
  • Offers 50 lb/in tensile strength
  • Solvent-resistant
  • Chemical-resistant
  • Provides crisp paint lines
  • Conformable
  • Tear-resistant
Catalog No. 50-007-92568 Supplier MSC Supplier No. 62472980
This masking tape is crafted from polyester film with a silicone adhesive, providing excellent performance for various applications. It measures 18 mm wide, 72 yd long, and 3.4 mil thick, featuring a distinctive green color.
  • Made from polyester film
  • Features silicone adhesive
  • Offers 50 lb/in tensile strength
  • Solvent-resistant
  • Chemical-resistant
  • Provides crisp paint lines
  • Conformable
  • Tear-resistant