Learn More
Enterprise Technology Solutions Intertape Polymer Group Tape, Med-Dty, Tamper-evident, 2.83x375', 8RL/CT, NL
SDP
Catalog No. 5000826303
Encompass_Preferred
Medium-duty tape adheres instantly to both virgin and recycled fiber surfaces to generate an immediate destructive bond. Starch-based adhesive aggressively bonds to corrugated cartons, even in dusty or dirty environments. Durable construction is not affected by extreme hot or cold temperatures to allow use in all weather conditions. Water-activated formulation leaves a broken seal for tamper-evident packages. Sealing tape is designed for use on lightweight to average-weight cartons up to 30 lb.
Catalog No. 50-008-26303 Supplier Enterprise Technology Solutions Supplier No. IPGK7004
Medium-duty tape adheres instantly to both virgin and recycled fiber surfaces to generate an immediate destructive bond. Starch-based adhesive aggressively bonds to corrugated cartons, even in dusty or dirty environments. Durable construction is not affected by extreme hot or cold temperatures to allow use in all weather conditions. Water-activated formulation leaves a broken seal for tamper-evident packages. Sealing tape is designed for use on lightweight to average-weight cartons up to 30 lb.