Learn More
MSC Intertape PVC Electrical Tape, 3/4in x 60ft, 1 ea
Catalog No. 5000685272
Encompass
This electrical insulating tape consists of a soft 'plasticized' stabilized PVC film coated with a non-corrosive rubber-based adhesive. It offers resistance to water, oils, acids, alkalis, and other corrosive chemicals.
  • Repairs frayed wiring
  • Splices electrical wiring
  • Protects wire-nut connections
  • Resurfaces and insulates hand tools
  • Suitable for general purpose maintenance and repair
  • Offers excellent adhesion and elongation properties
  • Provides good dielectric strength
  • Ideal for general insulation and bundling of cables
Catalog No. 50-006-85272 Supplier MSC Supplier No. 75416420
This electrical insulating tape consists of a soft 'plasticized' stabilized PVC film coated with a non-corrosive rubber-based adhesive. It offers resistance to water, oils, acids, alkalis, and other corrosive chemicals.
  • Repairs frayed wiring
  • Splices electrical wiring
  • Protects wire-nut connections
  • Resurfaces and insulates hand tools
  • Suitable for general purpose maintenance and repair
  • Offers excellent adhesion and elongation properties
  • Provides good dielectric strength
  • Ideal for general insulation and bundling of cables