Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

InterWorld Bouffant Cap 10070 Series

Catalog No. 191201194 Shop All Interworld Network Intl Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
21 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-120-1194 21 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-120-1194 Supplier InterWorld Supplier No. 1007010002
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

White polyethylene 21 in. bouffant cap protects workers and the environment.

  • General purpose bouffant cap
  • 21 in.
  • White
  • Polyethylene

Specifications

Product Type Bouffant
Size 21 in.
Material Polyethylene
Color White
Disposable Single-use
Size (English) 21 in.
Packaging Type Double Bagged
Diameter (English) 21 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.