Learn More
Medchemexpress LLC Intestinal Alkaline Phosphatase Antibody (YA1559) | 10 UL
SDP
Catalog No. 5000441708
Encompass_Preferred
The Intestinal Alkaline Phosphatase Antibody (YA1559) is a rabbit-derived, non-conjugated IgG monoclonal antibody designed to target Intestinal Alkaline Phosphatase. This enzyme is a key component of the gut mucosal defense system, functioning in the detoxification of lipopolysaccharide and preventing bacterial translocation within the gut. It can hydrolyze various phosphate compounds and is localized to the cell membrane as a homodimer.
  • Derived from rabbit
  • Monoclonal and recombinant
  • Non-conjugated
  • Reacts with human samples
  • Formulated in 50 mM Tris-Glycine (pH 7.4), 0.15 M NaCl, 40% Glycerol, 0.01% sodium azide and 0.05% BSA
  • Liquid appearance
  • Immunogen derived from human intestinal alkaline phosphatase
  • Ships with blue ice
Catalog No. 50-004-41708 Supplier Medchemexpress LLC Supplier No. HYP8181410UL
The Intestinal Alkaline Phosphatase Antibody (YA1559) is a rabbit-derived, non-conjugated IgG monoclonal antibody designed to target Intestinal Alkaline Phosphatase. This enzyme is a key component of the gut mucosal defense system, functioning in the detoxification of lipopolysaccharide and preventing bacterial translocation within the gut. It can hydrolyze various phosphate compounds and is localized to the cell membrane as a homodimer.
  • Derived from rabbit
  • Monoclonal and recombinant
  • Non-conjugated
  • Reacts with human samples
  • Formulated in 50 mM Tris-Glycine (pH 7.4), 0.15 M NaCl, 40% Glycerol, 0.01% sodium azide and 0.05% BSA
  • Liquid appearance
  • Immunogen derived from human intestinal alkaline phosphatase
  • Ships with blue ice