Learn More
Medchemexpress LLC Intestinal Alkaline Phosphatase Antibody (YA1559) | 50 UL
SDP
Catalog No. 5000441399
Encompass_Preferred
Intestinal Alkaline Phosphatase Antibody (YA1559) is a rabbit-derived, non-conjugated IgG monoclonal antibody that targets Intestinal Alkaline Phosphatase. This enzyme functions as an alkaline phosphatase, hydrolyzing various phosphate compounds. It is a component of the gut mucosal defense system, involved in the detoxification of lipopolysaccharide and the prevention of bacterial translocation in the gut.
  • Rabbit-derived, recombinant, monoclonal antibody
  • Reacts with human species
  • Immunogen is a synthesized peptide derived from human intestinal alkaline phosphatase aa1-55/528
  • Subcellular localization: cell membrane, lipid-anchor, GPI-anchor
  • Forms a homodimer
  • Supplied in 50mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% sodium azide and 0.05% BSA
  • For research use only
Catalog No. 50-004-41399 Supplier Medchemexpress LLC Supplier No. HYP8181450UL
Intestinal Alkaline Phosphatase Antibody (YA1559) is a rabbit-derived, non-conjugated IgG monoclonal antibody that targets Intestinal Alkaline Phosphatase. This enzyme functions as an alkaline phosphatase, hydrolyzing various phosphate compounds. It is a component of the gut mucosal defense system, involved in the detoxification of lipopolysaccharide and the prevention of bacterial translocation in the gut.
  • Rabbit-derived, recombinant, monoclonal antibody
  • Reacts with human species
  • Immunogen is a synthesized peptide derived from human intestinal alkaline phosphatase aa1-55/528
  • Subcellular localization: cell membrane, lipid-anchor, GPI-anchor
  • Forms a homodimer
  • Supplied in 50mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% sodium azide and 0.05% BSA
  • For research use only