Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human Integrin alpha 8 beta 1 (ITGA8&ITGB1) Heterodimer Protein;HEK293;200UG;IT1,Integrin alpha 8 beta 1,ITGA8&ITGB1,ITGA8&B1
Catalog No. 501657470
Encompass_Preferred
Recombinant Protein;Biotinylated Human Integrin alpha 8 beta 1 (ITGA8&ITGB1) Heterodimer Protein;200UG;Biotinylated Human ITGA8 & ITGB1 Heterodimer Protein (IT1-H82Wb) is expressed from human 293 cells (HEK293). It contains AA Phe 39 - Leu 1012 (ITGA8) & Gln 21 - Asp 728 (ITGB1) (Accession # P53708-1 (ITGA8) & P05556-1 (ITGB1)).;Biotinylated Human ITGA8 & ITGB1 Heterodimer Protein, produced by co-expression of ITGA8 and ITGB1, has a calculated MW of 116.4 kDa (ITGA8) & 83.7 kDa (ITGB1). Subunit ITGA8 is fused with an acidic tail at the C-terminus and followed by a polyhistidine tag and an Avi tag and subunit ITGB1 contains no tag but a basic tail at the C-terminus. The reducing (R) protein migrates as 110 kDa and 120 kDa due to glycosylation.;Immobilized Fibronectin at 5 μg/mL (100 μL/well) can bind Biotinylated Human ITGA8 & ITGB1 Heterodimer Protein (Cat. No. IT1-H82Wb) with a linear range of 0.156-5 μg/mL (QC tested).
Catalog No. 50-165-7470 Supplier ACROBiosystems Supplier No. IT1H82WB200UG
Recombinant Protein;Biotinylated Human Integrin alpha 8 beta 1 (ITGA8&ITGB1) Heterodimer Protein;200UG;Biotinylated Human ITGA8 & ITGB1 Heterodimer Protein (IT1-H82Wb) is expressed from human 293 cells (HEK293). It contains AA Phe 39 - Leu 1012 (ITGA8) & Gln 21 - Asp 728 (ITGB1) (Accession # P53708-1 (ITGA8) & P05556-1 (ITGB1)).;Biotinylated Human ITGA8 & ITGB1 Heterodimer Protein, produced by co-expression of ITGA8 and ITGB1, has a calculated MW of 116.4 kDa (ITGA8) & 83.7 kDa (ITGB1). Subunit ITGA8 is fused with an acidic tail at the C-terminus and followed by a polyhistidine tag and an Avi tag and subunit ITGB1 contains no tag but a basic tail at the C-terminus. The reducing (R) protein migrates as 110 kDa and 120 kDa due to glycosylation.;Immobilized Fibronectin at 5 μg/mL (100 μL/well) can bind Biotinylated Human ITGA8 & ITGB1 Heterodimer Protein (Cat. No. IT1-H82Wb) with a linear range of 0.156-5 μg/mL (QC tested).