Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human Integrin alpha V beta 5 (ITGAV&ITGB5) Heterodimer Protein;HEK293;200UG;IT5,Integrin alpha V beta 5,ITGAVB5,ITGAV&ITGB5
Supplier:  ACROBiosystems IT5H82WA200UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human Integrin alpha V beta 5 (ITGAV&ITGB5) Heterodimer Protein;200UG;Biotinylated Human ITGAV & ITGB5 Heterodimer Protein (IT5-H82Wa) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Val 992 (ITGAV) & Gly 24 - Asn 719 (ITGB5) (Accession # NP_002201.1 (ITGAV) & P18084-1 (ITGB5)).;Biotinylated Human ITGAV & ITGB5 Heterodimer Protein, produced by co-expression of ITGAV and ITGB5, has a calculated MW of 114.7 kDa (ITGAV) and 81.9 kDa (ITGB5). Subunit ITGAV is fused with an acidic tail at the C-terminus and followed by a polyhistidine tag and an Avi tag (Avitag™) and subunit ITGB5 contains no tag but a basic tail at the C-terminus. The reducing (R) protein migrates as 120 kDa and 150kDa (ITGAV) and 90 kDa (ITGB5) respectively due to glycosylation.;Immobilized Human Vitronectin at 5 μg/mL (100 μL/well) can bind Biotinylated Human ITGAV & ITGB5 Heterodimer Protein (Cat. No. IT5-H82Wa) with a linear range of 0.01-0.156 μg/mL
Catalog No. 50-165-7485