Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human Integrin alpha V beta 6 (ITGAV&ITGB6) Heterodimer Protein;HEK293;25UG;IT6,Integrin alpha V beta 6,ITGAV&ITGB6,Integrin,ITGAV,ITGA,ITGB6,ITGB,ITGAVB6
Supplier:  ACROBiosystems IT6H82E425UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human Integrin alpha V beta 6 (ITGAV&ITGB6) Heterodimer Protein;25UG;Biotinylated Human ITGAV&ITGB6, His Tag, Avi Tag (IT6-H82E4) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Val 992 (ITGAV) & Gly 22 - Asn 707 (ITGB6) (Accession # NP_002201.1 (ITGAV)& AAI21179.1 (ITGB6)).;Biotinylated Human ITGAV & ITGB6 Heterodimer Protein, produced by co-expression of ITGAV and ITGB6, has a calculated MW of 113 kDa (ITGAV) and 79.6 kDa (ITGB6). Subunit ITGAV is fused with an acidic tail at the C-terminus and followed by a polyhistidine tag and an Avi tag (Avitag™) and subunit ITGB6 contains no tag but a basic tail at the C-terminus. The predicted N-terminus is Phe 31 (ITGAV) & Gly 22 (ITGB6). The reducing (R) protein migrates as 100-140 kDa (ITGAV) and 145-160 kDa (ITGB6) respectively due to glycosylation.;Immobilized Human TGFB1, His Tag (Cat. No. TG1-H524x) at 5 μg/mL (100 μL/well) can bind Biotinylated Human ITGAV&ITGB6, His
Catalog No. 50-165-8194