Learn More
MSC Rockwell Labs Ltd Perimeter Bait, 10% Boric Acid, 10 lb, Granular
Catalog No. 502838402
Encompass
This granular perimeter bait effectively controls a variety of common pests. Containing 10% boric acid, it offers long-lasting, weather-resistant protection for indoor and outdoor applications, suitable for perimeter treatments, gardens, and food preparation areas.
  • Controls roaches, most ants, crickets, earwigs, silverfish, millipedes, sowbugs, pillbugs, mole crickets, snails, and slugs.
  • Exclusive 10% boric acid granular formula.
  • Economical outdoor application (1 lb per 1000 sq ft).
  • Non-repellent for colony elimination.
  • Weather-resistant, up to 90 days outdoor protection.
  • Approved for indoor (food areas) and outdoor use.
  • Mold and moisture resistant.
  • Mineral-based active ingredient.
Catalog No. 50-283-8402 Supplier MSC Supplier No. 74333170
This granular perimeter bait effectively controls a variety of common pests. Containing 10% boric acid, it offers long-lasting, weather-resistant protection for indoor and outdoor applications, suitable for perimeter treatments, gardens, and food preparation areas.
  • Controls roaches, most ants, crickets, earwigs, silverfish, millipedes, sowbugs, pillbugs, mole crickets, snails, and slugs.
  • Exclusive 10% boric acid granular formula.
  • Economical outdoor application (1 lb per 1000 sq ft).
  • Non-repellent for colony elimination.
  • Weather-resistant, up to 90 days outdoor protection.
  • Approved for indoor (food areas) and outdoor use.
  • Mold and moisture resistant.
  • Mineral-based active ingredient.