Learn More
Medchemexpress LLC Intoplicine dimesylate | 133711-99-6 | 98.3% | 540.65 | C23H32N4O7S2 | 50 MG
SDP
Catalog No. 5000249044
Encompass_Preferred
Intoplicine dimesylate is an antitumor derivative in the 7H-benzo[e]pyrido[4,3-b]indole series that inhibits DNA topoisomerase I and II and binds strongly to DNA, making it useful for biochemical and cellular studies of topoisomerase activity.
  • Dual topoisomerase I and II inhibitor.
  • Strong DNA binding (KA = 2 x 10^5 M^-1).
  • High reported purity suitable for research applications.
  • Solid form with distinct light blue to blue color for easy identification.
  • Stable under sealed storage; follow recommended low-temperature solvent storage for extended stability.
Catalog No. 50-002-49044 Supplier Medchemexpress LLC Supplier No. HY101647A50MG
Intoplicine dimesylate is an antitumor derivative in the 7H-benzo[e]pyrido[4,3-b]indole series that inhibits DNA topoisomerase I and II and binds strongly to DNA, making it useful for biochemical and cellular studies of topoisomerase activity.
  • Dual topoisomerase I and II inhibitor.
  • Strong DNA binding (KA = 2 x 10^5 M^-1).
  • High reported purity suitable for research applications.
  • Solid form with distinct light blue to blue color for easy identification.
  • Stable under sealed storage; follow recommended low-temperature solvent storage for extended stability.