Learn More
JANT PHARMACAL CORPORATION Intoxilyzer 900 - Automatic sampling breath alcohol instrument w/test storage, Bluetooth print capabilities. Includes Bluetooth Printer w/ Rechargeable Battery and charger, Carry Case, 2 Rolls paper, 10 Mouthpieces, Operator manual.
SDP
Catalog No. NC3457152
Encompass_Preferred
The Intoxilyzer 900 is the industry’s NEWEST portable handheld EBT device. The I-900 has a rugged ergonomic design that, when paired to a Bluetooth thermal printer, gives the BAT flexibility of testing in the workplace or on the go. A large bright multi-color display and easy to use menu makes the Intoxilyzer 900 effortless to use. PLUS, the it automatically calculates the corrected value of the barometric pressure for you during the calibration process. DOT, NRC, and DOE protocols are all built-in to the device software.
This item is not returnable. View return policy
Catalog No. NC3457152 Supplier JANT PHARMACAL CORPORATION Supplier No. DS900
The Intoxilyzer 900 is the industry’s NEWEST portable handheld EBT device. The I-900 has a rugged ergonomic design that, when paired to a Bluetooth thermal printer, gives the BAT flexibility of testing in the workplace or on the go. A large bright multi-color display and easy to use menu makes the Intoxilyzer 900 effortless to use. PLUS, the it automatically calculates the corrected value of the barometric pressure for you during the calibration process. DOT, NRC, and DOE protocols are all built-in to the device software.