Learn More
Creative Biomart Intra-Cellular Adhesion Molecule-1 (ICAM-1)
Catalog No. 502084302
Encompass_Preferred
ICAM-1 Human Recombinant is expressed in E. coli containing 455 amino acids 28-483 fused to an amino terminal hexahistidine tag. The ICAM-1 is purified by proprietary chromatographic techniques.
Catalog No. 50-208-4302 Supplier Creative Biomart Supplier No. THP0251
ICAM-1 Human Recombinant is expressed in E. coli containing 455 amino acids 28-483 fused to an amino terminal hexahistidine tag. The ICAM-1 is purified by proprietary chromatographic techniques.