Learn More
Perkin Elmer US LLC Intracooler Insulated Bolt Kit
Catalog No. 502112799
Encompass_Preferred
Intracoolers from PerkinElmer are subambient devices that automatically control the flow of coolant through a circulating chamber attached to the DSC 7, Pyres 1 DSC, and the Diamond DAC analyzers.
Catalog No. 50-211-2799 Supplier Perkin Elmer US LLC Supplier No. 03190247
Add to Cart
Add to Cart
Intracoolers from PerkinElmer are subambient devices that automatically control the flow of coolant through a circulating chamber attached to the DSC 7, Pyres 1 DSC, and the Diamond DAC analyzers.