Learn More
MOLECULAR DEPOT LLC Intralipid 20 - 1 mL
Catalog No. 5000013537
Encompass_Preferred
Intralipid for Interference Testing is a highly concentrated intralipid solution (20perc) used to test intralipids interference in various assays This product is for in vitro research use only and is not for use on humans For in vitro research use only
Catalog No. 50-000-13537 Supplier MOLECULAR DEPOT LLC Supplier No. K2010008I
Intralipid for Interference Testing is a highly concentrated intralipid solution (20perc) used to test intralipids interference in various assays This product is for in vitro research use only and is not for use on humans For in vitro research use only