Learn More
Med Vet International Intramedullary Steinman Pins, 1/16" x 7" Trocar/Trocar, J0250A
Catalog No. 5000583105
Encompass_Preferred
Highest quality type 316L stainless steel IM pins. Available in the points and diameters listed below. When ordering, please match diameter with point style number. Made in U.S.A.
Catalog No. 50-005-83105 Supplier Med Vet International Supplier No. JOR250A
Highest quality type 316L stainless steel IM pins. Available in the points and diameters listed below. When ordering, please match diameter with point style number. Made in U.S.A.