Learn More
3B Scientific™ Intramuscular Injection Simulator - Upper Arm
GREENER_CHOICESDPSCI_ED
Catalog No. S27329 Shop All 3B Scientific Products
Change view
Click to view available options
IM injection simulator - upper arm
Catalog No.
S27329 IM injection simulator - upper arm
1 options
Catalog No. S27329 Supplier 3B Scientific™ Supplier No. P55/1
Add to Cart
Add to Cart
3B Scientific™ Intramuscular Injection Simulator - Upper Arm features all important anatomical palpable landmarks such as acromion and humerus to allow for placing correct intramuscular injections in an graphic way.
  • Built-in fine electronics produce audio-visual feedback of the result, e.g. correct or incorrect injection, bone contact, or wrong location.
  • Additional trainings and control modes optionally provide an immediate or subsequent performance check under training conditions.
  • Included life-like silicone skin is extremely durable and can be quickly and easily exchanged when necessary.
  • Includes:
    • 2 AA batteries
    • Detailed instruction manual
    • Replacement skin
    • 5-mL injection syringe
    • 21G/0.8 injection needle
Specifications
Product Type IM Injection Simulator - Upper Arm
Includes 2 AA batteries, Instruction manual, Replacement skin, 5-mL injection syringe, 21G/0.8 injection needle
Material Silicone