Learn More
Cell Signaling Technology InTraSeq™ 3' Conjugate Antibody Cocktail 1, Human and Mouse Reactivity, 4 Assays
SDP
Catalog No. 503132868
Encompass
This antibody cocktail is a mixture of 31 titrated conjugates designed to measure proteins including cytoplasmic, surface, and nuclear targets, as well as post-translational modifications, alongside single-cell RNA sequencing. By enabling simultaneous protein and RNA measurement, this tool helps uncover cell states and biological data that RNA analysis alone may miss. This cocktail is validated for use with specialized single-cell sequencing technology to facilitate high-resolution cellular analysis.
  • Includes 31 titrated conjugates
  • Compatible with the specified single-cell assay kit
  • Validated for use with chromium single-cell kits using feature barcoding technology
  • Species reactivity covers human and mouse samples
  • Stable for at least 12 months when stored at -20°C
Catalog No. 50-313-2868 Supplier Cell Signaling Technology Supplier No. 48167S
This antibody cocktail is a mixture of 31 titrated conjugates designed to measure proteins including cytoplasmic, surface, and nuclear targets, as well as post-translational modifications, alongside single-cell RNA sequencing. By enabling simultaneous protein and RNA measurement, this tool helps uncover cell states and biological data that RNA analysis alone may miss. This cocktail is validated for use with specialized single-cell sequencing technology to facilitate high-resolution cellular analysis.
  • Includes 31 titrated conjugates
  • Compatible with the specified single-cell assay kit
  • Validated for use with chromium single-cell kits using feature barcoding technology
  • Species reactivity covers human and mouse samples
  • Stable for at least 12 months when stored at -20°C