Learn More
Braintree Scientific Intrathecal Catheter Alzet Pumps
SDP
Catalog No. 501955692
Encompass
Intrathecal Catheter - Braintree Scientific's rat intrathecal catheter offers a means for the administration of microliter amounts of drug solution to the lumbar (or higher) region of the rat spinal cord. This catheter system is ideal for use in antinociceptive studies involving rats. **Sold for research purposes only and not for human use**
Catalog No. 50-195-5692 Supplier Braintree Scientific Supplier No. RITC7743
Intrathecal Catheter - Braintree Scientific's rat intrathecal catheter offers a means for the administration of microliter amounts of drug solution to the lumbar (or higher) region of the rat spinal cord. This catheter system is ideal for use in antinociceptive studies involving rats. **Sold for research purposes only and not for human use**