Learn More
Braintree Scientific Intrathecal Catheter standard
SDP
Catalog No. 501955693
Encompass
Intrathecal Catheter - Braintree Scientific's rat intrathecal catheter offers a means for the administration of microliter amounts of drug solution to the lumbar (or higher) region of the rat spinal cord. This catheter system is ideal for use in antinociceptive studies involving rats. **Sold for research purposes only and not for human use**
Catalog No. 50-195-5693 Supplier Braintree Scientific Supplier No. RITCSTD
Intrathecal Catheter - Braintree Scientific's rat intrathecal catheter offers a means for the administration of microliter amounts of drug solution to the lumbar (or higher) region of the rat spinal cord. This catheter system is ideal for use in antinociceptive studies involving rats. **Sold for research purposes only and not for human use**