Learn More
SAI Infusion Technologies 50' Roll of PU-025 Tubing - 0.012" ID x 0.025" OD
SDP
Catalog No. NC9084371
Encompass
50 foot roll of PU-025 Tubing - 0.012" ID x 0.025" OD. SAI's Medical Grade Polyurethane micro-tubing is specially formulated for applications where intravascular thrombosis is a concern. It reduces blood platelet and other blood related deposits on the walls of the tubing, making it extremely blood compatible. While its initial stiffness makes it easy to implant into a vessel, it softens at body temperature for maximum biocompatibility. Product is for laboratory animal research only.
Catalog No. NC9084371 Supplier SAI Infusion Technologies Supplier No. PU02550
50 foot roll of PU-025 Tubing - 0.012" ID x 0.025" OD. SAI's Medical Grade Polyurethane micro-tubing is specially formulated for applications where intravascular thrombosis is a concern. It reduces blood platelet and other blood related deposits on the walls of the tubing, making it extremely blood compatible. While its initial stiffness makes it easy to implant into a vessel, it softens at body temperature for maximum biocompatibility. Product is for laboratory animal research only.