Learn More
SAI Infusion Technologies 50' Roll of 3F Silastic Tubing - 0.020" ID x 0.037" OD
SDP
Catalog No. NC1360594
Encompass
50 foot roll of 3F Medical Grade Silicone Tubing - 0.020" ID x 0.037" OD. SAI's Medical Grade Silicone micro-tubing is highly biocompatible and is resistant to chemicals. SAI's Platinum cured Silicone meets USDA Class VI standards and can be sterilized with Ethylene Oxide or autoclave. Product is for laboratory animal research only.
Catalog No. NC1360594 Supplier SAI Infusion Technologies Supplier No. SIL350
50 foot roll of 3F Medical Grade Silicone Tubing - 0.020" ID x 0.037" OD. SAI's Medical Grade Silicone micro-tubing is highly biocompatible and is resistant to chemicals. SAI's Platinum cured Silicone meets USDA Class VI standards and can be sterilized with Ethylene Oxide or autoclave. Product is for laboratory animal research only.