Learn More
SAI Infusion Technologies 50' Roll of PU-033 Tubing - 0.015" ID x 0.033" OD
SDP
Catalog No. NC0324347
Encompass
50 foot roll of PU-033 Tubing - 0.015" ID x 0.033" OD. SAI's Medical Grade Polyurethane micro-tubing is specially formulated for applications where intravascular thrombosis is a concern. It reduces blood platelet and other blood related deposits on the walls of the tubing, making it extremely blood compatible. While its initial stiffness makes it easy to implant into a vessel, it softens at body temperature for maximum biocompatibility. Product is for laboratory animal research only.
Catalog No. NC0324347 Supplier SAI Infusion Technologies Supplier No. PU03350
50 foot roll of PU-033 Tubing - 0.015" ID x 0.033" OD. SAI's Medical Grade Polyurethane micro-tubing is specially formulated for applications where intravascular thrombosis is a concern. It reduces blood platelet and other blood related deposits on the walls of the tubing, making it extremely blood compatible. While its initial stiffness makes it easy to implant into a vessel, it softens at body temperature for maximum biocompatibility. Product is for laboratory animal research only.