Learn More
Hanna Pharmaceutical Supply Co., Inc. Empty INTRAVIA Container 250ML
Catalog No. NC1234655
Encompass_Preferred
Due to recent natural disasters, production of sterile water has been negatively impacted.  Please expect long lead times on these itemsEmpty INTRAVIA Container 250 mL. Sterile, nonpyrogenic fluid path. Port Tubes, which contact fluid path are DEHP-plasticized PVC. 48/CS
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.
This item is not returnable. View return policy
Catalog No. NC1234655 Supplier Hanna Pharmaceutical Supply Co., Inc. Supplier No. BAX2B8012
Due to recent natural disasters, production of sterile water has been negatively impacted.  Please expect long lead times on these itemsEmpty INTRAVIA Container 250 mL. Sterile, nonpyrogenic fluid path. Port Tubes, which contact fluid path are DEHP-plasticized PVC. 48/CS
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.