Learn More
BioMed Diagnostics An enriched dermatophyte medium used in the detection of dermatophytes. | Detects: Trichophyton Mentagrophytes, T. rubrum, T. tonsurans, Microsporum gypseum. | Available in: 20 pack
Catalog No. NC2582843
Encompass
InTray Dermatophyte contains an enriched dermatophyte medium used to aid in the detection of dermatophytes from specimens with mixed micobiota. InTray Dermatophyte is formulated to produce a red color in the presence of growing dermatophytes. Moreover, the medium is formulated to assist in the macro-and microscopical identification of fungi characteristics. The medium inhibits most Gram-positive and Gram-negative bacteria, yeast and saprophytic fungi. It is a single exposure system with dynamic built-in components and features that are designed for user compatibility and ease of detection. | Detects: Trichophyton Mentagrophytes, T. rubrum, T. tonsurans, and Microsporum gypseum. | Time to Result: Incubate inoculated trays for up to 14 days at 18-30C, in the dark. Observe the trays daily for color changes, through the clear viewing window. | Available in: 20 pack
Catalog No. NC2582843 Supplier BioMed Diagnostics Supplier No. 12593002
Add to Cart
Add to Cart
InTray Dermatophyte contains an enriched dermatophyte medium used to aid in the detection of dermatophytes from specimens with mixed micobiota. InTray Dermatophyte is formulated to produce a red color in the presence of growing dermatophytes. Moreover, the medium is formulated to assist in the macro-and microscopical identification of fungi characteristics. The medium inhibits most Gram-positive and Gram-negative bacteria, yeast and saprophytic fungi. It is a single exposure system with dynamic built-in components and features that are designed for user compatibility and ease of detection. | Detects: Trichophyton Mentagrophytes, T. rubrum, T. tonsurans, and Microsporum gypseum. | Time to Result: Incubate inoculated trays for up to 14 days at 18-30C, in the dark. Observe the trays daily for color changes, through the clear viewing window. | Available in: 20 pack