Learn More
BioMed Diagnostics InTray Mueller Hinton Chocolate is for use in qualitative procedures for the isolation and cultivation of fastidious organisms. | Detects: Taylorella equigenitalis | Available in: 20 pack
Supplier:  BioMed Diagnostics 12483002
Encompass
Streamline your workflow with InTray Mueller Hinton Chocolate Agar. Whether used in the lab, or at the point-of-need, you're now able to combine the collection, transport, growth and resulting into one device. InTray Mueller Hinton Chocolate Agar is for use in qualitative procedures for the isolation and cultivation of fastidious organisms, particularly those of the Haemophilus species. | Detects: Taylorella equigenitalis | Time to Result: 18 - 72 hours at 37C | Available in: 20 pack
Catalog No. NC2582935