Learn More
LICOR Intercept T20 (PBS) Antibody Diluent (500 mL)
Catalog No. NC2031675
Encompass_Preferred
Intercept T20 (PBS) Antibody Diluent improves the specificity of the primary and secondary antibodies, reducing off-target effects.The diluent contains Intercept (PBS) Blocking Buffer preformulated with Tween 20 for use in a phosphate-buffered saline (PBS) system. Since there’s no need to mix the diluent yourself, it saves you time and reduces potential variation. Intercept T20 (PBS) Antibody Diluent contains a 0.05% concentration of Tween 20. It contains no sodium azide and is stored at 4 °C.
Catalog No. NC2031675 Supplier LICOR Supplier No. 92775001
Intercept T20 (PBS) Antibody Diluent improves the specificity of the primary and secondary antibodies, reducing off-target effects.The diluent contains Intercept (PBS) Blocking Buffer preformulated with Tween 20 for use in a phosphate-buffered saline (PBS) system. Since there’s no need to mix the diluent yourself, it saves you time and reduces potential variation. Intercept T20 (PBS) Antibody Diluent contains a 0.05% concentration of Tween 20. It contains no sodium azide and is stored at 4 °C.