Learn More
MOLECULAR DEPOT LLC Intrinsic Factor - 0.1 mg
Catalog No. 5000013544
Encompass_Preferred
Intrinsic Factor from Porcine Gastric Mucosa (0 1 mg) consists of the natural binder and transporter of Vitamin B12 purified from the porcine gastric mucosa This product has been used as a molecular tool in Vitamin B12 detection This is supplied as a solution For in vitro research use only
Catalog No. 50-000-13544 Supplier MOLECULAR DEPOT LLC Supplier No. B2010532
Intrinsic Factor from Porcine Gastric Mucosa (0 1 mg) consists of the natural binder and transporter of Vitamin B12 purified from the porcine gastric mucosa This product has been used as a molecular tool in Vitamin B12 detection This is supplied as a solution For in vitro research use only